This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TGM5 SIRNA Assays ›
Gene Symbol:
TGM5
Gene Name:
transglutaminase 5
Gene Aliases:
PSS2, TGASE5, TGASEX, TGM6, TGMX, TGX
Chromosome Location:
Chr.15: 43232595 - 43266857 on Build GRCh38
UniGene ID:
Hs.129719
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 14439

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TGM5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004245.3 NP_004236.1 2, 3 208
NM_201631.3 NP_963925.2 3, 4 454
GenBank mRNA AF035960.1 - 3, 4 454
AF035961.1 - 2, 3 208
BC119009.1 - 3, 4 454 40085374
BC122859.1 - 3, 4 454 40085372

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
epidermis development
peptide cross-linking
protein-glutamine gamma-glutamyltransferase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header