This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LIMK1 SIRNA Assays ›
Gene Symbol:
LIMK1
Gene Name:
LIM domain kinase 1
Gene Aliases:
LIMK, LIMK-1
Chromosome Location:
Chr.7: 74083777 - 74122525 on Build GRCh38
UniGene ID:
Hs.647035
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 143455

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LIMK1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204426.1 NP_001191355.1 6 765
NM_002314.3 NP_002305.1 7 987
GenBank mRNA AB209557.1 - 4 459
AF134379.1 - 7 773
AK026544.1 - 1 23
AK290162.1 - 8 912
AK300382.1 - 6 704
D26309.1 - 8 868

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
signal transduction
Rho protein signal transduction
nervous system development
actin cytoskeleton organization
positive regulation of actin filament bundle assembly
Fc-gamma receptor signaling pathway involved in phagocytosis
positive regulation of axon extension
negative regulation of ubiquitin-protein transferase activity
positive regulation of stress fiber assembly
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
zinc ion binding
heat shock protein binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0