This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM101B SIRNA Assays ›
Gene Symbol:
FAM101B
Gene Name:
family with sequence similarity 101 member B
Gene Aliases:
CFM1
Chromosome Location:
Chr.17: 439978 - 445940 on Build GRCh38
UniGene ID:
Hs.345588
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 141942

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM101B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182705.2 NP_874364.1 1, 2 386
GenBank mRNA BC031341.1 - 416 4908279

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

epithelial to mesenchymal transition
skeletal system morphogenesis
negative regulation of chondrocyte development
actin filament bundle organization
negative regulation of bone mineralization involved in bone maturation
filamin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header