This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SARNP SIRNA Assays ›
Gene Symbol:
SARNP
Gene Name:
SAP domain containing ribonucleoprotein
Gene Aliases:
CIP29, HCC1, HSPC316, THO1
Chromosome Location:
Chr.12: 55752463 - 55817756 on Build GRCh38
UniGene ID:
Hs.505676
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 141175

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SARNP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033082.3 NP_149073.1 6 380
NR_026722.1 5 333
NR_026723.1 6 380
GenBank mRNA AF161434.1 - 6 347
AF486281.1 - 6 325
AK222719.1 - 6 358
AK290508.1 - 6 348
BC007099.1 - 6 346 4273903
BC093051.1 - 5 299 30382893
BI520421.1 - 6 380

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025364, BC094393, AK003453, AK146248, AK088266, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
regulation of translation
mRNA 3'-end processing
DNA binding
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header