This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GINS3 SIRNA Assays ›
Gene Symbol:
GINS3
Gene Name:
GINS complex subunit 3
Gene Aliases:
PSF3
Chromosome Location:
Chr.16: 58392394 - 58406144 on Build GRCh38
UniGene ID:
Hs.47125
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 140730

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GINS3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126129.1 NP_001119601.1 4 1970
NM_001126130.1 NP_001119602.1 2 1619
NM_022770.3 NP_073607.2 3 1853
GenBank mRNA AK023974.1 - 2 1524
AK127454.1 - 1 2781
AL137379.1 - 1 1189
BC005879.1 - 3 1728 3529058
BC014437.1 - 3 1747 4870865

Back To Top

More Information


Gene Ontology Categories:

Function(s)

DNA strand elongation involved in DNA replication

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0