This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPR162 SIRNA Assays ›
Gene Symbol:
GPR162
Gene Name:
G protein-coupled receptor 162
Gene Aliases:
A-2, GRCA
Chromosome Location:
Chr.12: 6821797 - 6827418 on Build GRCh38
UniGene ID:
Hs.631654
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 140620

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GPR162
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014449.1 NP_055264.1 5 974
NM_019858.1 NP_062832.1 5 1976
GenBank mRNA AB209836.1 - 4 1383
BC002353.2 - 5 1068 2821031
BC007918.2 - 4 607
BC080585.1 - 5 955 3510624
U47925.1 - 5 974
U47928.1 - 5 1976

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0