This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM222B SIRNA Assays ›
Gene Symbol:
FAM222B
Gene Name:
family with sequence similarity 222 member B
Gene Aliases:
C17orf63
Chromosome Location:
Chr.17: 28755978 - 28855036 on Build GRCh38
UniGene ID:
Hs.564533
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 140530

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM222B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077498.2 NP_001070966.1 3 2622
NM_001288631.1 NP_001275560.1 4 2793
NM_001288632.1 NP_001275561.1 5 2704
NM_001288633.1 NP_001275562.1 4 2698
NM_001288634.1 NP_001275563.1 3 2691
NM_001288635.1 NP_001275564.1 4 2670
NM_001288636.1 NP_001275565.1 6 2694
NM_001288637.1 NP_001275566.1 5 2660
NM_001288638.1 NP_001275567.1 5 2592
NM_001288639.1 NP_001275568.1 4 2544
NM_001288640.1 NP_001275569.1 3 2510
NM_018182.3 NP_060652.2 4 2656
XM_011524986.2 XP_011523288.1 3 2587
XM_011524987.1 XP_011523289.1 4 2422
XM_011524988.2 XP_011523290.1 3 2638
XM_017024831.1 XP_016880320.1 4 2746
XM_017024832.1 XP_016880321.1 3 3011
XM_017024833.1 XP_016880322.1 3 2551
XM_017024834.1 XP_016880323.1 3 2749
XM_017024835.1 XP_016880324.1 4 2948
GenBank mRNA AK000147.1 - 1 1766
AK001562.1 - 4 2607
AK025980.1 - 1 1625
AK095569.1 - 3 2691
BC021228.2 - 3 2506 4040385
BX647766.1 - 3 2441

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0