This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KRT76 SIRNA Assays ›
Gene Symbol:
KRT76
Gene Name:
keratin 76
Gene Aliases:
HUMCYT2A, KRT2B, KRT2P
Chromosome Location:
Chr.12: 52768155 - 52777345 on Build GRCh38
UniGene ID:
Hs.654392
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 140285

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KRT76
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015848.4 NP_056932.2 9 1957
GenBank mRNA AK299390.1 - 9 1957
M99063.1 - 9 1934

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytoskeleton organization
epidermis development
sebaceous gland development
structural molecule activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0