This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MLEC SIRNA Assays ›
Gene Symbol:
MLEC
Gene Name:
malectin
Gene Aliases:
KIAA0152
Chromosome Location:
Chr.12: 120687124 - 120701864 on Build GRCh38
UniGene ID:
Hs.744910
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 140188

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MLEC
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303627.1 NP_001290556.1 5 4893
NM_001303628.1 NP_001290557.1 3 4941
NM_014730.3 NP_055545.1 5 5176
XM_011539031.1 XP_011537333.1 5 9591
XM_011539032.1 XP_011537334.1 6 9279
GenBank mRNA CR936818.1 - 2 4346
D63486.1 - 5 5133

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
protein folding
enzyme binding
carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0