This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPANK1 SIRNA Assays ›
Gene Symbol:
GPANK1
Gene Name:
G-patch domain and ankyrin repeats 1
Gene Aliases:
ANKRD59, BAT4, D6S54E, G5, GPATCH10
Chromosome Location:
Chr.6: 31661229 - 31666283 on Build GRCh38
UniGene ID:
Hs.247478
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 139919

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GPANK1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199237.1 NP_001186166.1 4 1728
NM_001199238.1 NP_001186167.1 4 1597
NM_001199239.1 NP_001186168.1 4 1594
NM_001199240.1 NP_001186169.1 4 1380
NM_033177.3 NP_149417.1 3 1704
XM_005249403.3 XP_005249460.1 4 1731
XM_006715204.1 XP_006715267.1 4 1409
XM_011514909.1 XP_011513211.1 4 1449
XM_011514910.1 XP_011513212.1 4 1446
GenBank mRNA AJ420471.1 - 3 1296
AK123707.1 - 4 1728
AK314056.1 - 4 1597
BC008783.1 - 3 1694 3629572
BC018875.2 - 3 1688

Back To Top

More Information


Gene Ontology Categories:

Process(es)

nucleic acid binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header