This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TIA1 SIRNA Assays ›
Gene Symbol:
TIA1
Gene Name:
TIA1 cytotoxic granule-associated RNA binding protein
Gene Aliases:
TIA-1, WDM
Chromosome Location:
Chr.2: 70209444 - 70248793 on Build GRCh38
UniGene ID:
Hs.413123
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 139892

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TIA1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022037.2 NP_071320.2 3 430
NM_022173.2 NP_071505.2 3 430
XM_005264527.1 XP_005264584.1 3 440
XM_005264528.4 XP_005264585.1 2 342
XM_005264531.1 XP_005264588.1 3 358
XM_011533077.1 XP_011531379.1 3 440
XM_011533078.1 XP_011531380.1 3 358
XM_011533081.2 XP_011531383.1 3 439
XM_017004789.1 XP_016860278.1 3 259
XM_017004790.1 XP_016860279.1 3 258
XM_017004791.1 XP_016860280.1 2 342
XM_017004792.1 XP_016860281.1 3 439
XM_017004793.1 XP_016860282.1 3 439
XM_017004794.1 XP_016860283.1 3 440
XM_017004795.1 XP_016860284.1 3 318
XM_017004796.1 XP_016860285.1 3 439
XM_017004797.1 XP_016860286.1 3 439
XM_017004798.1 XP_016860287.1 3 439
GenBank mRNA AB209211.1 - 3 441
AK303345.1 - 3 441
BC015944.1 - 3 237 3909177
CN295707.1 - 3 346
CX867570.1 - 3 403
DB136383.1 - 3 442
M77142.1 - 3 269

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
fibroblast growth factor receptor signaling pathway
negative regulation of translation
negative regulation of cytokine biosynthetic process
regulation of mRNA splicing, via spliceosome
protein localization to cytoplasmic stress granule
positive regulation of epithelial cell apoptotic process
nucleotide binding
protein binding
poly(A) binding
AU-rich element binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0