This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM193A SIRNA Assays ›
Gene Symbol:
FAM193A
Gene Name:
family with sequence similarity 193 member A
Gene Aliases:
C4orf8, RES4-22
Chromosome Location:
Chr.4: 2596288 - 2732575 on Build GRCh38
UniGene ID:
Hs.652364
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 139330

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM193A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256666.1 NP_001243595.1 7 891
NM_001256667.1 NP_001243596.1 8 963
NM_001256668.1 NP_001243597.1 7 891
NM_003704.3 NP_003695.3 7 891
NR_046335.1 7 891
NR_046336.1 7 891
XM_006713930.2 XP_006713993.2 8 954
XM_006713932.2 XP_006713995.2 9 1026
XM_011513590.1 XP_011511892.1 10 1104
XM_011513593.2 XP_011511895.1 7 825
XM_017008788.1 XP_016864277.1 9 947
GenBank mRNA AB000459.1 - 7 891
AB000460.1 - 7 891
AB000461.1 - 7 891
AB001563.1 - 3 810
AF040966.1 - 3 579
AK303732.1 - 3 463
AK308449.1 - 8 960
BC136646.1 - 7 861 9020636
BC144337.1 - 7 858
BC144339.1 - 7 861
BC144340.1 - 8 933 9052860
BM468825.1 - 5 510

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0