This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CUL1 SIRNA Assays ›
Gene Symbol:
CUL1
Gene Name:
cullin 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 148698178 - 148801110 on Build GRCh38
UniGene ID:
Hs.146806
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 139194

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CUL1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003592.2 NP_003583.2 9 1348
XM_005250060.4 XP_005250117.1 9 1384
XM_011516629.2 XP_011514931.1 9 1340
XM_011516630.2 XP_011514932.1 9 1241
XM_011516631.2 XP_011514933.1 9 1369
XM_011516632.2 XP_011514934.1 9 1267
XM_017012723.1 XP_016868212.1 8 1104
GenBank mRNA AF062536.1 - 8 1122
AK096163.1 - 10 1236
AK096505.1 - 2 1007
AK296909.1 - 8 1073
AK315023.1 - 9 1524
BC125119.1 - 9 1261 40118923
BC125120.1 - 9 1261 40118927
BX537409.1 - 9 1278
U58087.1 - 9 1050

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108627, XM_006236409, XM_006236411, XM_006236410, BC161932.
This siRNA also targets the following transcripts in Mouse: NM_012042, AF083216, AK147004, AK209075, BC029260, and others.

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
G2/M transition of mitotic cell cycle
protein polyubiquitination
stimulatory C-type lectin receptor signaling pathway
protein monoubiquitination
cell cycle arrest
cell proliferation
negative regulation of cell proliferation
organ morphogenesis
viral process
Wnt signaling pathway
protein ubiquitination
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
T cell receptor signaling pathway
stress-activated MAPK cascade
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
intrinsic apoptotic signaling pathway
ubiquitin-protein transferase activity
protein binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0