This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GDF3 SIRNA Assays ›
Gene Symbol:
GDF3
Gene Name:
growth differentiation factor 3
Gene Aliases:
KFS3, MCOP7, MCOPCB6
Chromosome Location:
Chr.12: 7689785 - 7695764 on Build GRCh38
UniGene ID:
Hs.86232
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 138550

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GDF3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020634.1 NP_065685.1 2 848
GenBank mRNA AF110641.1 - 2 823
AF263538.1 - 2 848
AY358875.1 - 2 877
BC030959.1 - 2 860 4610141

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
eye development
in utero embryonic development
response to dietary excess
endoderm development
mesoderm development
regulation of cell fate commitment
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of BMP signaling pathway
notochord development
somite rostral/caudal axis specification
growth
regulation of apoptotic process
regulation of MAPK cascade
negative regulation of epidermal cell differentiation
negative regulation of myoblast differentiation
cell development
formation of anatomical boundary
SMAD protein signal transduction
primitive streak formation
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0