This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM20B SIRNA Assays ›
Gene Symbol:
FAM20B
Gene Name:
family with sequence similarity 20 member B
Gene Aliases:
gxk1
Chromosome Location:
Chr.1: 179025804 - 179076574 on Build GRCh38
UniGene ID:
Hs.5737
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 138485

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM20B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324310.1 NP_001311239.1 2, 3 571
NM_001324311.1 NP_001311240.1 2, 3 581
NM_014864.3 NP_055679.1 2, 3 676
GenBank mRNA AB007944.1 - 2, 3 719
AB480690.1 - 2, 3 676
AK289989.1 - 2, 3 576
BC046441.1 - 2, 3 544 5197154
BC051794.1 - 2, 3 676 5492043

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

phosphorylation
positive regulation of protein targeting to mitochondrion
ATP binding
kinase activity
phosphotransferase activity, alcohol group as acceptor
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0