This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDK5R2 SIRNA Assays ›
Gene Symbol:
CDK5R2
Gene Name:
cyclin dependent kinase 5 regulatory subunit 2
Gene Aliases:
NCK5AI, P39, p39nck5ai
Chromosome Location:
Chr.2: 218959628 - 218962155 on Build GRCh38
UniGene ID:
Hs.158460
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 13838

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CDK5R2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003936.4 NP_003927.1 1 436
GenBank mRNA BC041771.1 - 1 409 4818320
BG703026.1 - 1 414
U34051.1 - 1 312

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cyclin-dependent protein serine/threonine kinase activity
neuron migration
cerebellum development
superior olivary nucleus maturation
hippocampus development
layer formation in cerebral cortex
positive regulation of calcium ion-dependent exocytosis
positive regulation of protein serine/threonine kinase activity
lipid binding
cyclin-dependent protein kinase 5 activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0