This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LAPTM4A SIRNA Assays ›
Gene Symbol:
LAPTM4A
Gene Name:
lysosomal protein transmembrane 4 alpha
Gene Aliases:
HUMORF13, LAPTM4, MBNT, Mtrp
Chromosome Location:
Chr.2: 20032650 - 20052028 on Build GRCh38
UniGene ID:
Hs.467807
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 138350

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LAPTM4A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014713.4 NP_055528.1 6, 7 1132
GenBank mRNA AK129875.1 - 6, 7 782
AY359028.1 - 6, 7 749
BC000421.2 - 6, 7 709 2964450
BC003158.1 - 6, 7 772 2958870
BC093075.1 - 6, 7 732 30402963
CR456758.1 - 6, 7 624
D14696.1 - 6, 7 770

Back To Top

More Information


Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s)

transport

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0