This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SPATA2 SIRNA Assays ›
Gene Symbol:
SPATA2
Gene Name:
spermatogenesis associated 2
Gene Aliases:
PD1, PPP1R145, tamo
Chromosome Location:
Chr.20: 49903391 - 49915543 on Build GRCh38
UniGene ID:
Hs.48513
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 138173

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SPATA2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135773.1 NP_001129245.1 3 702
NM_006038.3 NP_006029.1 3 618
XM_006723894.1 XP_006723957.1 3 538
XM_011529116.1 XP_011527418.1 3 575
GenBank mRNA AB018300.1 - 3 536
AK023229.1 - 3 573
AK289644.1 - 3 702
AK295536.1 - 2 180
BC009481.2 - 3 525 3953522
CB992178.1 - 3 633
U28164.1 - 3 583

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
spermatogenesis
cell differentiation
molecular_function
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0