This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VASH1 SIRNA Assays ›
Gene Symbol:
VASH1
Gene Name:
vasohibin 1
Gene Aliases:
KIAA1036
Chromosome Location:
Chr.14: 76761457 - 76783020 on Build GRCh38
UniGene ID:
Hs.525479
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 136648

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: VASH1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014909.4 NP_055724.1 7 2993
XM_017021087.1 XP_016876576.1 9 2068
XM_017021088.1 XP_016876577.1 8 2025
XM_017021089.1 XP_016876578.1 8 1884
XM_017021090.1 XP_016876579.1 7 1817
XM_017021091.1 XP_016876580.1 7 3493
GenBank mRNA AB028959.1 - 7 2448
AL832588.2 - 7 2696

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
negative regulation of endothelial cell proliferation
cell cycle arrest
response to wounding
negative regulation of endothelial cell migration
negative regulation of angiogenesis
negative regulation of blood vessel endothelial cell migration
negative regulation of lymphangiogenesis
regulation of cellular senescence
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0