This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KERA SIRNA Assays ›
Gene Symbol:
KERA
Gene Name:
keratocan
Gene Aliases:
CNA2, KTN, SLRR2B
Chromosome Location:
Chr.12: 91050491 - 91058354 on Build GRCh38
UniGene ID:
Hs.125750
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 136191

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KERA
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007035.3 NP_008966.1 2 950
GenBank mRNA AF063301.1 - 1 339
AF205403.1 - 2 579
AK314990.1 - 2 512
BC032667.2 - 1 340 5541591
BM722433.1 - 2 483
CV571245.1 - 2 497

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

axonogenesis
visual perception
keratan sulfate biosynthetic process
keratan sulfate catabolic process
response to stimulus
cornea development in camera-type eye
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0