This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HMGN4 SIRNA Assays ›
Gene Symbol:
HMGN4
Gene Name:
high mobility group nucleosomal binding domain 4
Gene Aliases:
HMG17L3, NHC
Chromosome Location:
Chr.6: 26538344 - 26546937 on Build GRCh38
UniGene ID:
Hs.236774
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 135687

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HMGN4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006353.2 NP_006344.1 2 939
XM_017010171.1 XP_016865660.1 2 998
XM_017010172.1 XP_016865661.1 3 1062
XM_017010173.1 XP_016865662.1 4 1100
GenBank mRNA BC001282.1 - 2 883 3452950
BC035998.1 - 1 362
U90549.1 - 1 1392

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding
nucleosomal DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0