This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRDN SIRNA Assays ›
Gene Symbol:
TRDN
Gene Name:
triadin
Gene Aliases:
CPVT5, TDN, TRISK
Chromosome Location:
Chr.6: 123216339 - 123637093 on Build GRCh38
UniGene ID:
Hs.144744
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 135591

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRDN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006073.3 NP_006064.2 30 2018
GenBank mRNA CR749591.1 - 19 726
U18985.1 - 30 1722

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular calcium ion homeostasis
muscle contraction
regulation of cell communication by electrical coupling
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
cytoplasmic microtubule organization
ion transmembrane transport
regulation of release of sequestered calcium ion into cytosol
heart contraction
negative regulation of ryanodine-sensitive calcium-release channel activity
positive regulation of ryanodine-sensitive calcium-release channel activity
regulation of cardiac muscle cell membrane potential
endoplasmic reticulum membrane organization
positive regulation of cell communication by electrical coupling involved in cardiac conduction
regulation of cardiac conduction
receptor binding
protein binding
protein binding, bridging
ion channel binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0