This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other COPS7A SIRNA Assays ›
Gene Symbol:
COPS7A
Gene Name:
COP9 signalosome subunit 7A
Gene Aliases:
CSN7, CSN7A, SGN7a
Chromosome Location:
Chr.12: 6723984 - 6731875 on Build GRCh38
UniGene ID:
Hs.530823
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 134969

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: COPS7A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164093.1 NP_001157565.1 7 1936
NM_001164094.1 NP_001157566.1 8 1801
NM_001164095.2 NP_001157567.1 8 1872
NM_016319.3 NP_057403.1 8 1774
XM_005253694.2 XP_005253751.1 8 1699
GenBank mRNA AF193844.1 - 7 1562
AF210052.1 - 8 1703
AK001318.1 - 8 1686
AK021721.1 - 8 1739
AK022488.1 - 8 1681
AK128792.1 - 6 1471
BC011789.2 - 8 1662 4025790
BC093015.1 - 8 1327 30344094

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA damage recognition
transcription-coupled nucleotide-excision repair
cullin deneddylation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0