This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD207 SIRNA Assays ›
Gene Symbol:
CD207
Gene Name:
CD207 molecule
Gene Aliases:
CLEC4K
Chromosome Location:
Chr.2: 70825248 - 70860787 on Build GRCh38
UniGene ID:
Hs.199731
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 134671

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CD207
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015717.4 NP_056532.4 6 1015
XM_011532874.1 XP_011531176.1 7 1203
GenBank mRNA AJ242859.1 - 6 1015
AK314927.1 - 6 1009
BC022278.1 - 6 1008 4692155

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
receptor-mediated endocytosis
defense response to virus
mannose binding
carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0