This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPR160 SIRNA Assays ›
Gene Symbol:
GPR160
Gene Name:
G protein-coupled receptor 160
Gene Aliases:
GPCR1, GPCR150
Chromosome Location:
Chr.3: 170037323 - 170085415 on Build GRCh38
UniGene ID:
Hs.231320
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 134282

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GPR160
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014373.2 NP_055188.1 4 1225
XM_005247346.3 XP_005247403.1 5 1835
XM_005247347.3 XP_005247404.1 3 871
XM_006713582.3 XP_006713645.1 4 1187
XM_011512678.2 XP_011510980.1 5 1517
XM_017006161.1 XP_016861650.1 5 1328
XM_017006162.1 XP_016861651.1 5 1332
XM_017006163.1 XP_016861652.1 4 1849
GenBank mRNA AJ249248.1 - 3 956
BC000181.2 - 4 1160 3048193

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0