This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM184B SIRNA Assays ›
Gene Symbol:
TMEM184B
Gene Name:
transmembrane protein 184B
Gene Aliases:
C22orf5, FM08, HS5O6A, HSPC256
Chromosome Location:
Chr.22: 38216045 - 38273034 on Build GRCh38
UniGene ID:
Hs.182626
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 133885

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM184B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195071.1 NP_001182000.1 4, 5 668
NM_001195072.1 NP_001182001.1 5, 6 754
NM_012264.4 NP_036396.2 4, 5 653
XM_011530112.2 XP_011528414.2 4, 5 702
XM_011530113.1 XP_011528415.1 4, 5 632
XM_011530114.1 XP_011528416.1 4, 5 664
XM_011530115.2 XP_011528417.1 3, 4 423
XM_011530116.2 XP_011528418.2 4, 5 694
XM_011530117.1 XP_011528419.1 5, 6 730
XM_011530118.1 XP_011528420.1 5, 6 704
XM_011530119.1 XP_011528421.1 3, 4 458
XM_017028754.1 XP_016884243.1 4, 5 703
XM_017028755.1 XP_016884244.1 4, 5 696
XM_017028756.1 XP_016884245.1 4, 5 632
XM_017028757.1 XP_016884246.1 3, 4 420
XM_017028758.1 XP_016884247.1 4, 5 632
XM_017028759.1 XP_016884248.1 3, 4 458
GenBank mRNA AB097053.1 - 4, 5 624
AK075417.1 - 2, 3 141
AK292652.1 - 4, 5 653
AK309071.1 - 4, 5 689
AL096879.1 - 3, 4 346
BC015489.1 - 4, 5 609 3896682
BI197128.1 - 3, 4 347
BX648345.1 - 4, 5 704
CR456413.1 - 3, 4 487

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0