This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF123 SIRNA Assays ›
Gene Symbol:
RNF123
Gene Name:
ring finger protein 123
Gene Aliases:
FP1477, KPC1
Chromosome Location:
Chr.3: 49688753 - 49721529 on Build GRCh38
UniGene ID:
Hs.553723
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 133486

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF123
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022064.4 NP_071347.2 14 1271
NR_135218.1 14 1271
XM_006713285.1 XP_006713348.1 14 1429
XM_011533995.1 XP_011532297.1 14 1206
XM_017007018.1 XP_016862507.1 14 1233
GenBank mRNA AF370367.1 - 5 426
AK090632.1 - 4 314
AK096353.1 - 14 1185
AK308130.1 - 14 1213
AK308607.1 - 11 949
AL832034.1 - 14 1233
AL832138.1 - 14 1208
AL832344.1 - 14 1210
AY744152.1 - 13 1154
BC130632.1 - 14 1212 40146663
CR627051.1 - 12 1779

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
zinc ion binding
ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0