This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRC8A SIRNA Assays ›
Gene Symbol:
LRRC8A
Gene Name:
leucine rich repeat containing 8 family member A
Gene Aliases:
AGM5, LRRC8, SWELL1
Chromosome Location:
Chr.9: 128882112 - 128918042 on Build GRCh38
UniGene ID:
Hs.643600
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 132883

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRRC8A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127244.1 NP_001120716.1 4 4561
NM_001127245.1 NP_001120717.1 3 4185
NM_019594.3 NP_062540.2 4 4292
XM_005252095.2 XP_005252152.1 4 4422
XM_005252096.3 XP_005252153.1 4 5585
XM_006717186.2 XP_006717249.1 5 4637
XM_011518863.1 XP_011517165.1 4 4228
XM_011518864.2 XP_011517166.1 5 4346
XM_011518865.2 XP_011517167.1 3 4064
GenBank mRNA AB037858.1 - 3 4101
AY358286.1 - 2 3326

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

pre-B cell differentiation
anion transport
cell volume homeostasis
response to osmotic stress
regulation of anion transport
anion transmembrane transport
volume-sensitive anion channel activity
anion channel activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0