This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DNAJA4 SIRNA Assays ›
Gene Symbol:
DNAJA4
Gene Name:
DnaJ heat shock protein family (Hsp40) member A4
Gene Aliases:
MST104, MSTP104, PRO1472
Chromosome Location:
Chr.15: 78264145 - 78282196 on Build GRCh38
UniGene ID:
Hs.513053
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 132581

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DNAJA4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130182.1 NP_001123654.1 5 955
NM_001130183.1 NP_001123655.1 5 782
NM_018602.3 NP_061072.3 6 1094
XM_005254535.3 XP_005254592.1 6 867
XM_006720601.1 XP_006720664.1 6 869
GenBank mRNA AF173388.1 - 1 693
AK090898.1 - 2 426
AK096616.1 - 5 955
AK098079.1 - 6 1085
BC021720.2 - 6 1094 4831235
BC031044.1 - 5 817 5277834
BX648710.1 - 6 976

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to heat
protein refolding
negative regulation of inclusion body assembly
protein binding
ATP binding
heat shock protein binding
metal ion binding
unfolded protein binding
chaperone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0