This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZBTB14 SIRNA Assays ›
Gene Symbol:
ZBTB14
Gene Name:
zinc finger and BTB domain containing 14
Gene Aliases:
ZF5, ZFP-161, ZFP-5, ZFP161, ZNF478
Chromosome Location:
Chr.18: 5289019 - 5297053 on Build GRCh38
UniGene ID:
Hs.592340
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 13214

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZBTB14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143823.2 NP_001137295.1 4 1410
NM_001243702.1 NP_001230631.1 4 1078
NM_001243704.1 NP_001230633.1 4 1069
NM_003409.4 NP_003400.2 3 1380
GenBank mRNA AK313236.1 - 3 1223
BC057282.1 - 4 1256
BC110519.2 - 2 964 40033453
D89859.1 - 2 985
Y12726.1 - 2 952

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
heart valve development
cardiac septum development
transcription, DNA-templated
negative regulation of transcription, DNA-templated
coronary vasculature development
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0