This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC39A13 SIRNA Assays ›
Gene Symbol:
SLC39A13
Gene Name:
solute carrier family 39 member 13
Gene Aliases:
LZT-Hs9
Chromosome Location:
Chr.11: 47407132 - 47416500 on Build GRCh38
UniGene ID:
Hs.523664
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 131812

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SLC39A13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128225.2 NP_001121697.1 5, 6 822
NM_152264.4 NP_689477.2 5, 6 843
NR_134854.1 5, 6 912
XM_006718381.3 XP_006718444.1 5, 6 727
XM_006718384.2 XP_006718447.1 5, 6 727
XM_011520466.1 XP_011518768.1 5, 6 727
XM_011520467.1 XP_011518769.1 5, 6 1056
XM_011520468.2 XP_011518770.1 5, 6 1149
XM_017018540.1 XP_016874029.1 5, 6 701
XM_017018542.1 XP_016874031.1 5, 6 701
GenBank mRNA AK098651.1 - 5, 6 912
BC008853.2 - 5, 6 688 4100291
BC019016.2 - 5, 6 707 4100057
BI918365.1 - 4, 5 404
BX342307.2 - 5, 6 821
BX647491.1 - 7, 8 1007
CF132216.1 - 4, 5 429

Back To Top

More Information


Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

cellular zinc ion homeostasis
connective tissue development
zinc II ion transmembrane transport
zinc ion transmembrane transporter activity
protein binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0