This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PHF6 SIRNA Assays ›
Gene Symbol:
PHF6
Gene Name:
PHD finger protein 6
Gene Aliases:
BFLS, BORJ, CENP-31
Chromosome Location:
Chr.X: 134373312 - 134428792 on Build GRCh38
UniGene ID:
Hs.356501
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 131344

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PHF6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015877.1 NP_001015877.1 3, 4 439
NM_032335.3 NP_115711.2 3, 4 439
NM_032458.2 NP_115834.1 3, 4 439
GenBank mRNA AB058726.1 - 3, 4 325
AK290095.1 - 3, 4 410
AL832725.1 - 3, 4 432
AY157622.1 - 3, 4 439
BC005994.1 - 3, 4 378 4064169
CD685336.1 - 3, 4 447
DR000282.1 - 3, 4 474

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
zinc ion binding
tubulin binding
enzyme binding
histone binding
histone deacetylase binding
ribonucleoprotein complex binding
poly(A) RNA binding
phosphoprotein binding
scaffold protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header