This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRADC1 SIRNA Assays ›
Gene Symbol:
PRADC1
Gene Name:
protease associated domain containing 1
Gene Aliases:
C2orf7, PAP21
Chromosome Location:
Chr.2: 73228006 - 73233228 on Build GRCh38
UniGene ID:
Hs.61268
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 131276

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PRADC1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032319.1 NP_115695.1 1, 2 142
GenBank mRNA AK312077.1 - 1, 2 150
AY358605.1 - 1, 2 133
BC005069.1 - 1, 2 142 3530601

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0