This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRC4 SIRNA Assays ›
Gene Symbol:
LRRC4
Gene Name:
leucine rich repeat containing 4
Gene Aliases:
NAG14, NGL-2
Chromosome Location:
Chr.7: 128027071 - 128032273 on Build GRCh38
UniGene ID:
Hs.655003
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 130302

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRRC4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022143.4 NP_071426.1 2 2054
XM_011516461.2 XP_011514763.1 3 2360
XM_017012505.1 XP_016867994.1 3 2250
GenBank mRNA AF196976.4 - 2 2057
AJ297858.1 - 2 2045
AK172751.1 - 2 2058
AK314047.1 - 2 2130
AY358307.1 - 1 1973
BC111561.1 - 2 2048 40068142
BC111745.1 - 2 2048 40068146

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001037336, DQ119102.
This siRNA also targets the following transcripts in Mouse: NM_138682, AF300458, AK046781, BC012209, DQ177325, and others.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
cytokine-mediated signaling pathway
negative regulation of JAK-STAT cascade
regulation of synapse organization
postsynaptic density protein 95 clustering
protein kinase inhibitor activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0