This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF169 SIRNA Assays ›
Gene Symbol:
ZNF169
Gene Name:
zinc finger protein 169
Gene Aliases:
-
Chromosome Location:
Chr.9: 94259266 - 94303967 on Build GRCh38
UniGene ID:
Hs.387623
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 130150

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF169
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301275.1 NP_001288204.1 5 1141
NM_003448.2 NP_003439.2 5 777
NM_194320.3 NP_919301.2 5 774
XM_017014362.1 XP_016869851.1 5 906
XM_017014363.1 XP_016869852.1 6 800
XM_017014364.1 XP_016869853.1 6 797
XM_017014365.1 XP_016869854.1 5 7130
XM_017014366.1 XP_016869855.1 5 7068
GenBank mRNA AK093956.1 - 5 1079
AK122832.1 - 1 1397
AK289742.1 - 5 718
BC047702.2 - 4 698 5585128

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0