This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM26D SIRNA Assays ›
Gene Symbol:
FAM26D
Gene Name:
family with sequence similarity 26 member D
Gene Aliases:
C6orf78
Chromosome Location:
Chr.6: 116529013 - 116561123 on Build GRCh38
UniGene ID:
Hs.350750
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 129614

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM26D
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256887.2 NP_001243816.1 4 1421
NM_001256888.2 NP_001243817.1 4 1327
NM_001256889.2 NP_001243818.1 3 1205
NM_153036.4 NP_694581.1 3 1260
XM_005266860.3 XP_005266917.1 2 2195
XM_011535560.2 XP_011533862.1 5 1515
XM_011535561.2 XP_011533863.1 4 1364
XM_011535562.2 XP_011533864.1 6 1696
XM_011535563.2 XP_011533865.1 4 1309
XM_011535564.1 XP_011533866.1 4 1270
XM_017010390.1 XP_016865879.1 5 1443
XM_017010391.1 XP_016865880.1 5 1398
XM_017010392.1 XP_016865881.1 5 1460
GenBank mRNA AK056801.1 - 3 1193
AY358252.1 - 3 1242

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ion transmembrane transport
cation transmembrane transport
cation channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0