This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C9ORF43 SIRNA Assays ›
Gene Symbol:
C9orf43
Gene Name:
chromosome 9 open reading frame 43
Gene Aliases:
-
Chromosome Location:
Chr.9: 113410644 - 113429684 on Build GRCh38
UniGene ID:
Hs.632691
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 129579

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C9orf43
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278629.1 NP_001265558.1 7 903
NM_001278630.1 NP_001265559.1 7 760
NM_152786.2 NP_689999.1 7 976
XM_005251885.3 XP_005251942.1 5 282
XM_011518477.1 XP_011516779.1 7 903
XM_011518478.1 XP_011516780.1 7 903
XM_011518479.2 XP_011516781.1 5 558
GenBank mRNA AK301872.1 - 7 760
AK313867.1 - 7 892
BC026884.1 - 7 942 3455891
BC033037.2 - 7 861 4827930

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0