This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLHL40 SIRNA Assays ›
Gene Symbol:
KLHL40
Gene Name:
kelch like family member 40
Gene Aliases:
KBTBD5, NEM8, SRYP, SYRP
Chromosome Location:
Chr.3: 42685519 - 42692446 on Build GRCh38
UniGene ID:
Hs.350288
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 129443

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KLHL40
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152393.3 NP_689606.2 1 1204
GenBank mRNA AK056577.1 - 1 1186
AK291294.1 - 1 1186
AK300682.1 - 2 510
AY176040.1 - 1 1204
AY177390.1 - 1 1204
BC110491.2 - 1 1172 40007554

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
negative regulation of protein ubiquitination
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
skeletal muscle fiber development
ubiquitin-protein transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0