This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM210A SIRNA Assays ›
Gene Symbol:
FAM210A
Gene Name:
family with sequence similarity 210 member A
Gene Aliases:
C18orf19, HsT2329
Chromosome Location:
Chr.18: 13663347 - 13726592 on Build GRCh38
UniGene ID:
Hs.13034
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 129436

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM210A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098801.1 NP_001092271.1 3 643
NM_152352.3 NP_689565.2 2 503
GenBank mRNA AK055618.1 - 3 599
BU189118.1 - 1 90
DA895646.1 - 2 473
DA984075.1 - 2 500

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header