This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CSMD2 SIRNA Assays ›
Gene Symbol:
CSMD2
Gene Name:
CUB and Sushi multiple domains 2
Gene Aliases:
dJ1007G16.1, dJ1007G16.2, dJ947L8.1
Chromosome Location:
Chr.1: 33513999 - 34165842 on Build GRCh38
UniGene ID:
Hs.656915
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 128854

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CSMD2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281956.1 NP_001268885.1 4 755
NM_052896.4 NP_443128.2 4 487
XM_017000185.1 XP_016855674.1 4 749
XM_017000186.1 XP_016855675.1 4 749
XM_017000187.1 XP_016855676.1 4 578
XM_017000188.1 XP_016855677.1 4 487
XM_017000189.1 XP_016855678.1 4 749
XM_017000190.1 XP_016855679.1 4 749
XM_017000191.1 XP_016855680.1 4 749
XM_017000192.1 XP_016855681.1 4 749
XM_017000193.1 XP_016855682.1 4 749
XM_017000194.1 XP_016855683.1 4 487
GenBank mRNA AB212622.1 - 4 755
AY210418.1 - 4 487

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header