This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PASD1 SIRNA Assays ›
Gene Symbol:
PASD1
Gene Name:
PAS domain containing 1
Gene Aliases:
CT63, OXTES1
Chromosome Location:
Chr.X: 151563535 - 151676739 on Build GRCh38
UniGene ID:
Hs.160594
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 128217

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PASD1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173493.2 NP_775764.2 11 1210
XM_011531102.1 XP_011529404.1 11 1123
GenBank mRNA AK097552.1 - 11 1123
AY270020.1 - 11 1016
BC040301.1 - 11 1021 4826573

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of circadian rhythm
negative regulation of transcription, DNA-templated
rhythmic process
transcription coactivator binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0