This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MROH2B SIRNA Assays ›
Gene Symbol:
MROH2B
Gene Name:
maestro heat like repeat family member 2B
Gene Aliases:
HEATR7B2
Chromosome Location:
Chr.5: 40998020 - 41071342 on Build GRCh38
UniGene ID:
Hs.97714
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 128212

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MROH2B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173489.4 NP_775760.3 4, 5 851
XM_011513952.1 XP_011512254.1 4, 5 837
XM_011513953.1 XP_011512255.1 3, 4 775
GenBank mRNA BC034600.1 - 4, 5 815
BC052962.1 - 4, 5 812 4838071

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008760805.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0