This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM155 SIRNA Assays ›
Gene Symbol:
TMEM155
Gene Name:
transmembrane protein 155
Gene Aliases:
-
Chromosome Location:
Chr.4: 121758930 - 121765400 on Build GRCh38
UniGene ID:
Hs.27524
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 127957

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM155
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317837.1 NP_001304766.1 6 1172
NM_001317838.1 NP_001304767.1 6 1169
NM_001317839.1 NP_001304768.1 6 1047
NM_001317841.1 NP_001304770.1 5 1462
NM_001317842.1 NP_001304771.1 5 1306
NM_152399.3 NP_689612.3 6 1328
XM_005262737.2 XP_005262794.1 6 1136
XM_017007725.1 XP_016863214.1 6 1042
XM_017007726.1 XP_016863215.1 5 982
GenBank mRNA AK055396.1 - 6 1049
BC047657.1 - 6 1328 5264721

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0