This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHMP4C SIRNA Assays ›
Gene Symbol:
CHMP4C
Gene Name:
charged multivesicular body protein 4C
Gene Aliases:
SNF7-3, Shax3, VPS32C
Chromosome Location:
Chr.8: 81732453 - 81759515 on Build GRCh38
UniGene ID:
Hs.183861
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 127954

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CHMP4C
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152284.3 NP_689497.1 1 350
GenBank mRNA AB120734.1 - 1 182
AK000049.1 - 1 313
AK314873.1 - 1 349
AY329086.1 - 1 184
BC014321.1 - 1 354 3828410
BX492886.1 - 1 370

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell separation after cytokinesis
autophagy
nucleus organization
mitotic metaphase plate congression
abscission
regulation of centrosome duplication
protein transport
endosomal transport
viral life cycle
negative regulation of cytokinesis
multivesicular body assembly
viral budding via host ESCRT complex
regulation of viral process
ubiquitin-independent protein catabolic process via the multivesicular body sorting pathway
regulation of mitotic spindle assembly
positive regulation of viral release from host cell
protein binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0