This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM110B SIRNA Assays ›
Gene Symbol:
FAM110B
Gene Name:
family with sequence similarity 110 member B
Gene Aliases:
C8orf72
Chromosome Location:
Chr.8: 57994444 - 58149718 on Build GRCh38
UniGene ID:
Hs.154652
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 127937

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM110B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147189.2 NP_671722.1 5 1178
XM_005251324.2 XP_005251381.1 4 1172
XM_005251325.3 XP_005251382.1 3 11335
XM_005251326.2 XP_005251383.1 3 11360
XM_017013948.1 XP_016869437.1 4 3747
GenBank mRNA AK026141.1 - 2 712
AK092221.1 - 5 1178
AY937246.1 - 1 410
BC024294.1 - 2 690 5440447
DQ431182.1 - 1 298
U79298.1 - 1 309

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0