This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNX1 SIRNA Assays ›
Gene Symbol:
SNX1
Gene Name:
sorting nexin 1
Gene Aliases:
HsT17379, VPS5
Chromosome Location:
Chr.15: 64094071 - 64144234 on Build GRCh38
UniGene ID:
Hs.188634
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 12783

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SNX1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242933.1 NP_001229862.1 7, 8 854
NM_003099.4 NP_003090.2 7, 8 854
NM_148955.3 NP_683758.1 5, 6 659
XM_005254611.2 XP_005254668.1 7, 8 830
GenBank mRNA AB209013.1 - 7, 8 661
AF065483.1 - 7, 8 724
AF065484.1 - 5, 6 529
AK128179.1 - 7, 8 746
AK222561.1 - 7, 8 738
AK222793.1 - 7, 8 727
AK291752.1 - 7, 8 738
AK310618.1 - 4, 5 495
BC000357.2 - 7, 8 734 2964409
BQ935515.1 - 7, 8 728
BT006983.1 - 7, 8 724
U53225.1 - 7, 8 769

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
vesicle organization
receptor internalization
early endosome to Golgi transport
retrograde transport, endosome to Golgi
positive regulation of protein catabolic process
lamellipodium morphogenesis
cell-cell adhesion
epidermal growth factor receptor binding
insulin receptor binding
protein binding
phosphatidylinositol binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
cadherin binding involved in cell-cell adhesion
transferrin receptor binding
leptin receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0