This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM78A SIRNA Assays ›
Gene Symbol:
FAM78A
Gene Name:
family with sequence similarity 78 member A
Gene Aliases:
C9orf59
Chromosome Location:
Chr.9: 131258078 - 131281930 on Build GRCh38
UniGene ID:
Hs.143878
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 127731

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM78A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033387.3 NP_203745.2 2 2884
XM_011518567.1 XP_011516869.1 3 2427
XM_011518568.2 XP_011516870.1 2 3130
GenBank mRNA AK024434.1 - 3 3506
BC029924.2 - 2 2298
BC049388.2 - 2 2884 6050655

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0