This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF674 SIRNA Assays ›
Gene Symbol:
ZNF674 KRBOX4
Gene Name:
zinc finger protein 674
KRAB box domain containing 4
Gene Aliases:
MRX92, ZNF673B
ZNF673
Chromosome Location:
Chr.X: 46497725 - 46545459 on Build GRCh38
Chromosome Location:
Chr.X: 46447189 - 46474639 on Build GRCh38
UniGene ID:
Hs.636105
Hs.632800
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 127007

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF674
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039891.2 NP_001034980.1 6 508
NM_001146291.1 NP_001139763.1 6 490
NM_001190417.1 NP_001177346.1 6 493
XM_011543941.2 XP_011542243.1 6 505
XM_011543943.2 XP_011542245.1 6 507
XM_011543944.2 XP_011542246.1 4 315
XM_011543945.2 XP_011542247.1 3 264
XM_017029728.1 XP_016885217.1 7 660
XM_017029729.1 XP_016885218.1 7 682
XM_017029730.1 XP_016885219.1 7 679
XM_017029731.1 XP_016885220.1 3 264
XM_017029732.1 XP_016885221.1 7 657
XM_017029733.1 XP_016885222.1 7 659
GenBank mRNA AK295054.1 - 6 422
AY971607.1 - 6 492
DC317540.1 - 6 477
Gene Symbol: KRBOX4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129898.1 NP_001123370.1 6 580
NM_001129899.1 NP_001123371.1 7 624
NM_001129900.1 NP_001123372.1 7 609
NM_017776.2 NP_060246.2 6 565
GenBank mRNA AK000351.1 - 6 430
AK096413.1 - 6 472
AK097159.1 - 1 1454
AK289703.1 - 6 462
BC012569.1 - 6 432 4283420
DA152034.1 - 7 555
DA631861.1 - 7 526

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding
nucleic acid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0