This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NSA2 SIRNA Assays ›
Gene Symbol:
NSA2
Gene Name:
NSA2, ribosome biogenesis homolog
Gene Aliases:
CDK105, HCL-G1, HCLG1, HUSSY-29, HUSSY29, TINP1
Chromosome Location:
Chr.5: 74765429 - 74776912 on Build GRCh38
UniGene ID:
Hs.482526
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 126785

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NSA2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271665.1 NP_001258594.1 3 660
NM_014886.4 NP_055701.1 3 660
NR_073403.1 3 660
XM_011543098.1 XP_011541400.1 3 338
XM_017008952.1 XP_016864441.1 3 338
XM_017008953.1 XP_016864442.1 3 410
GenBank mRNA AF077615.1 - 3 374
AF372458.1 - 3 385
AJ012409.1 - 3 372
AK313784.1 - 3 403
AY443419.1 - 2 290
BC005288.1 - 3 403 3683720
CB052084.1 - 1 24

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

maturation of 5.8S rRNA
maturation of LSU-rRNA
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0