This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C3ORF67 SIRNA Assays ›
Gene Symbol:
C3orf67
Gene Name:
chromosome 3 open reading frame 67
Gene Aliases:
-
Chromosome Location:
Chr.3: 58716415 - 59050078 on Build GRCh38
UniGene ID:
Hs.368434
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 126589

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C3orf67
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198463.2 NP_940865.1 6 526
XM_005264929.2 XP_005264986.1 6 906
XM_006713013.3 XP_006713076.1 6 906
XM_011533451.2 XP_011531753.1 6 906
XM_011533453.2 XP_011531755.1 6 906
XM_011533455.2 XP_011531757.1 6 906
XM_011533456.1 XP_011531758.1 6 615
XM_011533457.1 XP_011531759.1 4 395
XM_011533458.2 XP_011531760.1 3 278
XM_011533459.2 XP_011531761.1 6 906
XM_011533460.2 XP_011531762.1 1 35
GenBank mRNA AK124111.1 - 6 528
AK296111.1 - 6 620
BC050317.1 - 6 893
BC132815.1 - 4 340 40146837
BC140002.1 - 7 636
BC151142.1 - 4 340 9020498

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0